accessibility menu, dialog, popup

UserName

Viewing profile as user:
HCOOH

CAS RN 30964-00-2

IUPAC Name: hept-6-ynoic acid
Synonyms: 6-heptynoic acid
Molecular Weight (g/mol): 126.16
Molecular Formula: C7H10O2
InChi Key: OFCPMJGTZUVUSM-UHFFFAOYSA-N
SMILES: OC(=O)CCCCC#C
Molecular Weight (g/mol) 
  • (1)
  • (6)
Percent Purity 
  • (1)
  • (6)
Quantity 
  • (3)
  • (2)
  • (2)
Boiling Point 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (1)
  • (6)
  • (1)
  • (6)
  • (3)
  • (2)
  • (2)
  • (4)