accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HOO

CAS RN 3098-18-8

IUPAC Name: 2-benzoyl-5-methylphenol
Synonyms: 2-benzoyl-5-methylphenol
Molecular Weight (g/mol): 212.25
Molecular Formula: C14H12O2
InChi Key: KIZCNUWGIVQQBK-UHFFFAOYSA-N
SMILES: CC1=CC=C(C(=O)C2=CC=CC=C2)C(O)=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results