accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OOIrO

CAS RN 30980-84-8

IUPAC Name: dioxoiridium hydrate
Synonyms: dioxoiridium hydrate
Molecular Weight (g/mol): 242.23
Molecular Formula: H2IrO3
InChi Key: YEPXCFHWKVNVAX-UHFFFAOYSA-N
SMILES: O.O=[Ir]=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)