accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHOCH3

CAS RN 30981-98-7

IUPAC Name: 2-(2,3-dimethylphenyl)acetic acid
Synonyms: (2,3-dimethylphenyl)acetic acid
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: AMPNBNGNPLXMOP-UHFFFAOYSA-N
SMILES: CC1=C(C)C(CC(O)=O)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)