accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClN

CAS RN 3099-28-3

IUPAC Name: 2,6-bis(chloromethyl)pyridine
Synonyms: 2,6-bis(chloromethyl)pyridine
Molecular Weight (g/mol): 176.04
Molecular Formula: C7H7Cl2N
InChi Key: IWQNFYRJSVJWQA-UHFFFAOYSA-N
SMILES: ClCC1=CC=CC(CCl)=N1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)