accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOOOH3C

CAS RN 31008-19-2

IUPAC Name: 5-[4-(3,4-dimethoxyphenyl)-hexahydrofuro[3,4-c]furan-1-yl]-2H-1,3-benzodioxole
Synonyms: 5-[4-(3,4-dimethoxyphenyl)-hexahydrofuro[3,4-c]furan-1-yl]-2H-1,3-benzodioxole
Molecular Weight (g/mol): 370.4
Molecular Formula: C21H22O6
InChi Key: AWOGQCSIVCQXBT-UHFFFAOYNA-N
SMILES: COC1=C(OC)C=C(C=C1)C1OCC2C1COC2C1=CC2=C(OCO2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results