accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 31008-64-7

IUPAC Name: trilithium(1+) ({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Synonyms: trilithium(1+) adenosine 5'-diphosphate
Molecular Weight (g/mol): 445
Molecular Formula: C10H12Li3N5O10P2
InChi Key: LZGPPAHUZSOGHJ-DJXXCMMGNA-K
SMILES: [Li+].[Li+].[Li+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
Physical Form 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)