accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3NH2CH3H3CH2N

CAS RN 3102-87-2

IUPAC Name: tetramethylbenzene-1,4-diamine
Synonyms: tetramethylbenzene-1,4-diamine
Molecular Weight (g/mol): 164.25
Molecular Formula: C10H16N2
InChi Key: WCZNKVPCIFMXEQ-UHFFFAOYSA-N
SMILES: CC1=C(C)C(N)=C(C)C(C)=C1N
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)