accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 31024-26-7

IUPAC Name: (3-aminopropyl)(methoxy)dimethylsilane
Synonyms: (3-aminopropyl)(methoxy)dimethylsilane
Molecular Weight (g/mol): 147.29
Molecular Formula: C6H17NOSi
InChi Key: MCLXOMWIZZCOCA-UHFFFAOYSA-N
SMILES: CO[Si](C)(C)CCCN
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)