accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 311-46-6

IUPAC Name: ethyl 2-(dimethoxyphosphoryl)acetate
Synonyms: ethyl 2-(dimethoxyphosphoryl)acetate
Molecular Weight (g/mol): 196.14
Molecular Formula: C6H13O5P
InChi Key: HUNISAHOCCASGM-UHFFFAOYSA-N
SMILES: CCOC(=O)CP(=O)(OC)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results