accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3114-55-4

IUPAC Name: 1-chloro(²H₅)benzene
Synonyms: 1-chloro(²H₅)benzene
Molecular Weight (g/mol): 117.59
Molecular Formula: C6H5Cl
InChi Key: MVPPADPHJFYWMZ-RALIUCGRSA-N
SMILES: [2H]C1=C([2H])C([2H])=C(Cl)C([2H])=C1[2H]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results