accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3115-68-2

IUPAC Name: tetrabutylphosphanium bromide
Synonyms: tetrabutylphosphonium bromide
Molecular Weight (g/mol): 339.34
Molecular Formula: C16H36BrP
InChi Key: RKHXQBLJXBGEKF-UHFFFAOYSA-M
SMILES: [Br-].CCCC[P+](CCCC)(CCCC)CCCC
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (5)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)