CAS RN 3117-03-1
IUPAC Name:
2,5-dimethoxycyclohexa-2,5-diene-1,4-dione
Synonyms:
2,5-dimethoxycyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol):
168.15
Molecular Formula:
C8H8O4
InChi Key:
RMMPZDDLWLALLJ-UHFFFAOYSA-N
SMILES:
COC1=CC(=O)C(OC)=CC1=O
1
–
1
of
1
results
1
2,5-Dimethoxy-1,4-benzoquinone 98.0+%, TCI America™
CAS: 3117-03-1 Molecular Formula: C8H8O4 Molecular Weight (g/mol): 168.15 MDL Number: MFCD00189386 InChI Key: RMMPZDDLWLALLJ-UHFFFAOYSA-N Synonym: 2,5-Dimethoxy-p-quinone PubChem CID: 101405 IUPAC Name: 2,5-dimethoxycyclohexa-2,5-diene-1,4-dione SMILES: COC1=CC(=O)C(=CC1=O)OC