accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSOHH3C

CAS RN 3120-74-9

IUPAC Name: 3-methyl-4-(methylsulfanyl)phenol
Synonyms: MMTP methylthiomethylphenol usaf MA-17
Molecular Weight (g/mol): 154.23
Molecular Formula: C8H10OS
InChi Key: VKALYYFVKBXHTF-UHFFFAOYSA-N
SMILES: CSC1=CC=C(O)C=C1C
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)