accessibility menu, dialog, popup

UserName

Viewing profile as user:
INCS

CAS RN 3125-73-3

IUPAC Name: 1-iodo-3-isothiocyanatobenzene
Synonyms: 1-iodo-3-isothiocyanatobenzene
Molecular Weight (g/mol): 261.08
Molecular Formula: C7H4INS
InChi Key: LKCSXVAACHUTGC-UHFFFAOYSA-N
SMILES: IC1=CC=CC(=C1)N=C=S
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results