accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 312624-26-3

CH2NZnBr

CAS RN 312624-26-3

IUPAC Name: 2-cyanoethanide; bromozincylium
Synonyms: 2-cyanoethanide; bromozincylium
Molecular Weight (g/mol): 199.36
Molecular Formula: C3H4BrNZn
InChi Key: CREYONULNRWHIX-UHFFFAOYSA-M
SMILES: [Zn+]Br.[CH2-]CC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results