accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNHNOHCl

CAS RN 312636-16-1

IUPAC Name: 4-{[4-(4-chlorophenyl)-1,3-thiazol-2-yl]amino}phenol
Synonyms: 4-{[4-(4-chlorophenyl)-1,3-thiazol-2-yl]amino}phenol
Molecular Weight (g/mol): 302.78
Molecular Formula: C15H11ClN2OS
InChi Key: ZFGXZJKLOFCECI-UHFFFAOYSA-N
SMILES: OC1=CC=C(NC2=NC(=CS2)C2=CC=C(Cl)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results