accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H2CH3CZnBr

CAS RN 312693-02-0

IUPAC Name: 2-ethylbutan-1-ide; bromozincylium
Synonyms: 2-ethylbutan-1-ide; bromozincylium
Molecular Weight (g/mol): 230.45
Molecular Formula: C6H13BrZn
InChi Key: VSNDAARQTRCDDM-UHFFFAOYSA-M
SMILES: [Zn+]Br.CCC([CH2-])CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results