accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 312693-05-3

CH2FClZn

CAS RN 312693-05-3

IUPAC Name: zinc(2+) (2-fluorophenyl)methanide chloride
Synonyms: zinc(2+) (2-fluorophenyl)methanide chloride
Molecular Weight (g/mol): 209.95
Molecular Formula: C7H6ClFZn
InChi Key: PLNYKKXFDVJFHW-UHFFFAOYSA-M
SMILES: [Cl-].[Zn++].[CH2-]C1=CC=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results