CAS RN 312693-07-5
IUPAC Name:
(4-fluorophenyl)methanide; chlorozincylium
Synonyms:
(4-fluorophenyl)methanide; chlorozincylium
Molecular Weight (g/mol):
209.95
Molecular Formula:
C7H6ClFZn
InChi Key:
HEXMIUITECMJJV-UHFFFAOYSA-M
SMILES:
Cl[Zn+].[CH2-]C1=CC=C(F)C=C1
1
–
1
of
1
results
1
Sigma Aldrich Bismuth(III) Phosphate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.