accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3127-14-8

IUPAC Name: 4,4,4',4'-tetramethyl-3,3',4,4'-tetrahydro-2,2'-spirobi[[1]benzopyran]-7,7'-diol
Synonyms: 4,4,4',4'-tetramethyl-3H,3'H-2,2'-spirobi[[1]benzopyran]-7,7'-diol
Molecular Weight (g/mol): 340.42
Molecular Formula: C21H24O4
InChi Key: BWQOXWHQZXVXSD-UHFFFAOYSA-N
SMILES: CC1(C)CC2(CC(C)(C)C3=C(O2)C=C(O)C=C3)OC2=C1C=CC(O)=C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results