accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSOON

CAS RN 31271-90-6

IUPAC Name: 1-(4-methylbenzenesulfonyl)-1H-indole
Synonyms: 1-(4-methylbenzenesulfonyl)indole
Molecular Weight (g/mol): 271.33
Molecular Formula: C15H13NO2S
InChi Key: JNRRPYFLDADLJW-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)N1C=CC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results