accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3128-06-1

IUPAC Name: 5-oxohexanoic acid
Synonyms: 5-oxohexanoic acid 5-keto-n-caproic acid delta-oxocaproic acid delta-ketocaproic acid
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: MGTZCLMLSSAXLD-UHFFFAOYSA-N
SMILES: CC(=O)CCCC(O)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results