accessibility menu, dialog, popup

UserName

Viewing profile as user:
FePH3CCH3CH3H3CCH3CH3PhPhPhPhPh

CAS RN 312959-24-3

IUPAC Name: 1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene
Synonyms:
Molecular Weight (g/mol): 710.72
Molecular Formula: C48H47FeP
InChi Key:
SMILES: [Fe].CC(C)(C)P(c1cccc1)C(C)(C)C.C1=CC=C(C=C1)c1c(c(c(c1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)