accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2H2NOOHO

CAS RN 3130-87-8

IUPAC Name: 2-amino-3-carbamoylpropanoic acid
Synonyms: asparagin asparagina Hasp
Molecular Weight (g/mol): 132.12
Molecular Formula: C4H8N2O3
InChi Key: DCXYFEDJOCDNAF-UHFFFAOYNA-N
SMILES: NC(CC(N)=O)C(O)=O
Percent Purity 
  • (4)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)