accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 31301-28-7

IUPAC Name: 4-methyl-1,10-phenanthroline
Synonyms: 4-methyl-1,10-phenanthroline
Molecular Weight (g/mol): 194.24
Molecular Formula: C13H10N2
InChi Key: NAZZKEZTSOOCSZ-UHFFFAOYSA-N
SMILES: CC1=C2C=CC3=CC=CN=C3C2=NC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results