accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3131-52-0

IUPAC Name: 1H-indole-5,6-diol
Synonyms: dihydroxyindole DHI dopamine lutine
Molecular Weight (g/mol): 149.15
Molecular Formula: C8H7NO2
InChi Key: SGNZYJXNUURYCH-UHFFFAOYSA-N
SMILES: OC1=C(O)C=C2C=CNC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results