accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrO

CAS RN 3132-64-7

IUPAC Name: 2-(bromomethyl)oxirane
Synonyms: epibromohydrin epibromhidrina epibromohydrine epibromhydrine α-epibromohydrin epibromhydrin
Molecular Weight (g/mol): 136.98
Molecular Formula: C3H5BrO
InChi Key: GKIPXFAANLTWBM-UHFFFAOYNA-N
SMILES: BrCC1CO1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)