accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 313545-31-2

IUPAC Name: (4-ethoxy-2-methylphenyl)boronic acid
Synonyms: 4-ethoxy-2-methylphenylboronic acid
Molecular Weight (g/mol): 180.01
Molecular Formula: C9H13BO3
InChi Key: MBEQXBKSJCOKJF-UHFFFAOYSA-N
SMILES: CCOC1=CC(C)=C(C=C1)B(O)O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results