CAS RN 313667-46-8
IUPAC Name:
N-(3-chloro-4-methylphenyl)-2-methoxybenzamide
Synonyms:
N-(3-chloro-4-methylphenyl)-2-methoxybenzamide
Molecular Weight (g/mol):
275.73
Molecular Formula:
C15H14ClNO2
InChi Key:
SASNELOZVTWLSU-UHFFFAOYSA-N
SMILES:
COC1=CC=CC=C1C(=O)NC1=CC(Cl)=C(C)C=C1
1
–
1
of
1
results
1
N-(3-Chloro-4-methylphenyl)-2-methoxybenzamide, 97%, Thermo Scientific™
CAS: 313667-46-8 Molecular Formula: C15H14ClNO2 Molecular Weight (g/mol): 275.732 MDL Number: MFCD00702336 InChI Key: SASNELOZVTWLSU-UHFFFAOYSA-N Synonym: n-3-chloro-4-methylphenyl-2-methoxybenzamide,cambridge id 5303656 PubChem CID: 2189574 IUPAC Name: N-(3-chloro-4-methylphenyl)-2-methoxybenzamide SMILES: CC1=C(C=C(C=C1)NC(=O)C2=CC=CC=C2OC)Cl