CAS RN 314-40-9
CAS RN 314-40-9
IUPAC Name:
5-bromo-3-(butan-2-yl)-6-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms:
bromacil
Molecular Weight (g/mol):
261.12
Molecular Formula:
C9H13BrN2O2
InChi Key:
CTSLUCNDVMMDHG-UHFFFAOYNA-N
SMILES:
CCC(C)N1C(=O)NC(C)=C(Br)C1=O
1
–
1
of
1
results
1
Bromacil, SPEX CertiPrep™
CAS: 314-40-9 Molecular Formula: C9H13BrN2O2 Molecular Weight (g/mol): 261.119 InChI Key: CTSLUCNDVMMDHG-UHFFFAOYSA-N PubChem CID: 9411 ChEBI: CHEBI:83658 IUPAC Name: 5-bromo-3-butan-2-yl-6-methyl-1H-pyrimidine-2,4-dione SMILES: CCC(C)N1C(=O)C(=C(NC1=O)C)Br