accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 314054-21-2

IUPAC Name: 2,6-difluoro-N-(2-phenylethyl)benzamide
Synonyms: 2,6-difluoro-N-(2-phenylethyl)benzamide
Molecular Weight (g/mol): 261.27
Molecular Formula: C15H13F2NO
InChi Key: PORBEPWKQBWEGX-UHFFFAOYSA-N
SMILES: FC1=CC=CC(F)=C1C(=O)NCCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results