accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 31410-01-2

IUPAC Name: 1-(prop-2-en-1-yl)-1H-imidazole
Synonyms: 1-(prop-2-en-1-yl)imidazole
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: XLXCHZCQTCBUOX-UHFFFAOYSA-N
SMILES: C=CCN1C=CN=C1
Boiling Point 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)