CAS RN 314291-83-3
CAS RN 314291-83-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ PTP Inhibitor V, PHPS1
The PTP Inhibitor V, PHPS1, also referenced under CAS 314291-83-3, controls the biological activity of PTP. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.