CAS RN 31430-41-8
IUPAC Name:
3-chloro-5-methylpyridin-2-amine
Synonyms:
3-chloro-5-methylpyridin-2-amine
Molecular Weight (g/mol):
142.59
Molecular Formula:
C6H7ClN2
InChi Key:
COTHZSSUQJRBQH-UHFFFAOYSA-N
SMILES:
CC1=CC(Cl)=C(N)N=C1
1
–
1
of
1
results
1
2-Amino-3-chloro-5-methylpyridine 98.0+%, TCI America™
CAS: 31430-41-8 Molecular Formula: C6H7ClN2 Molecular Weight (g/mol): 142.586 InChI Key: COTHZSSUQJRBQH-UHFFFAOYSA-N Synonym: 6-Amino-5-chloro-3-picoline PubChem CID: 12251011 IUPAC Name: 3-chloro-5-methylpyridin-2-amine SMILES: CC1=CC(=C(N=C1)N)Cl