CAS RN 31437-20-4
IUPAC Name:
(pyrimidin-2-yl)thiourea
Synonyms:
pyrimidin-2-ylthiourea
Molecular Weight (g/mol):
154.19
Molecular Formula:
C5H6N4S
InChi Key:
MCCLFFQZNBEOAV-UHFFFAOYSA-N
SMILES:
NC(=S)NC1=NC=CC=N1