accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3144-16-9

IUPAC Name: [(1R,4S)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonate
Synonyms: (S)-camphorsulfonate
Molecular Weight (g/mol): 231.29
Molecular Formula: C10H15O4S
InChi Key: MIOPJNTWMNEORI-XVKPBYJWSA-M
SMILES: CC1(C)[C@H]2CC[C@]1(CS([O-])(=O)=O)C(=O)C2
Molecular Weight (g/mol) 
  • (8)
  • (5)
Quantity 
  • (5)
  • (2)
  • (1)
  • (5)
Percent Purity 
  • (3)
  • (3)
  • (2)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results