accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOSiCH3CH3CH3H3CH3C

CAS RN 31469-15-5

IUPAC Name: [(1-methoxy-2-methylprop-1-en-1-yl)oxy]trimethylsilane
Synonyms: [(1-methoxy-2-methylprop-1-en-1-yl)oxy]trimethylsilane
Molecular Weight (g/mol): 174.32
Molecular Formula: C8H18O2Si
InChi Key: JNOGVQJEBGEKMG-UHFFFAOYSA-N
SMILES: COC(O[Si](C)(C)C)=C(C)C
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)