accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3147-39-5

IUPAC Name: methyl 2,4,6-trihydroxybenzoate
Synonyms: methyl 2,4,6-trihydroxybenzoate
Molecular Weight (g/mol): 184.15
Molecular Formula: C8H8O5
InChi Key: AQDIJIAUYXOCGX-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(O)C=C(O)C=C1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results