accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NOOHHO

CAS RN 3147-45-3

IUPAC Name: 2,4-dihydroxybenzamide
Synonyms: 2,4-dihydroxybenzamide
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: IIUJCQYKTGNRHH-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=C(O)C=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results