CAS RN 3147-76-0
IUPAC Name:
2-(2H-1,2,3-benzotriazol-2-yl)-4-tert-butylphenol
Synonyms:
2-(1,2,3-benzotriazol-2-yl)-4-tert-butylphenol
Molecular Weight (g/mol):
267.33
Molecular Formula:
C16H17N3O
InChi Key:
WXHVQMGINBSVAY-UHFFFAOYSA-N
SMILES:
CC(C)(C)C1=CC=C(O)C(=C1)N1N=C2C=CC=CC2=N1
1
–
1
of
1
results
1
2-(5-tert-Butyl-2-hydroxyphenyl)benzotriazole 98.0+%, TCI America™
CAS: 3147-76-0 Molecular Formula: C16H17N3O Molecular Weight (g/mol): 267.332 MDL Number: MFCD03844795 InChI Key: WXHVQMGINBSVAY-UHFFFAOYSA-N PubChem CID: 76605 IUPAC Name: 2-(benzotriazol-2-yl)-4-tert-butylphenol SMILES: CC(C)(C)C1=CC(=C(C=C1)O)N2N=C3C=CC=CC3=N2