accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 315-12-8

IUPAC Name: 5-fluoro-1,3-dimethyl-2-nitrobenzene
Synonyms: 5-fluoro-1,3-dimethyl-2-nitrobenzene
Molecular Weight (g/mol): 169.16
Molecular Formula: C8H8FNO2
InChi Key: KDPQDERPJHLWGF-UHFFFAOYSA-N
SMILES: CC1=CC(F)=CC(C)=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results