accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 315-14-0

IUPAC Name: 1,3,5-trifluoro-2-nitrobenzene
Synonyms: 1,3,5-trifluoro-2-nitrobenzene
Molecular Weight (g/mol): 177.08
Molecular Formula: C6H2F3NO2
InChi Key: PWRFDGYYJWQIAB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(F)C=C(F)C=C1F
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)