accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 315-31-1

IUPAC Name: 2-fluoro-3-methylbenzoic acid
Synonyms: 2-fluoro-3-methylbenzoic acid
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: DGNAETGARNTCIL-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C(O)=O)=C1F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)