accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO(R)O(S)ONOO(R)OH(S)HO(R)HO

CAS RN 3150-24-1

IUPAC Name: (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
Synonyms: p-nitrophenyl-β-D-galactoside 4-nitrophenyl-β-D-galactoside 4-nitrophenylgalactoside
Molecular Weight (g/mol): 301.25
Molecular Formula: C12H15NO8
InChi Key: IFBHRQDFSNCLOZ-YBXAARCKSA-N
SMILES: OC[C@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@H]1O
Quantity 
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Grade 
  • (2)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)