accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH3C(Z)

CAS RN 31501-11-8

IUPAC Name: (3Z)-hex-3-en-1-yl hexanoate
Synonyms: cis-3-hexenyl hexanoate
Molecular Weight (g/mol): 198.31
Molecular Formula: C12H22O2
InChi Key: RGACQXBDYBCJCY-ALCCZGGFSA-N
SMILES: CCCCCC(=O)OCC\C=C/CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results