accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 31502-14-4

IUPAC Name: (2E)-non-2-en-1-ol
Synonyms: (2E)-non-2-en-1-ol
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: NSSALFVIQPAIQK-BQYQJAHWSA-N
SMILES: CCCCCC\C=C\CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results