accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 315203-26-0

IUPAC Name: bis(4-bromophenyl)but-2-enedinitrile
Synonyms: bis(4-bromophenyl)but-2-enedinitrile
Molecular Weight (g/mol): 388.06
Molecular Formula: C16H8Br2N2
InChi Key: RWHQRUSYDYNSTI-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C(C#N)=C(C#N)C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results