accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHOH

CAS RN 31566-31-1

IUPAC Name: 2,3-dihydroxypropyl octadecanoate
Synonyms: glyceryl stearate grocor 5500 emcol msk lipo gms 600 dermagine lipo gms 450 lipo gms 410 drewmulse TP monostearate (glyceride) abracol S.L.G.
Molecular Weight (g/mol): 358.56
Molecular Formula: C21H42O4
InChi Key: VBICKXHEKHSIBG-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)OCC(O)CO
Molecular Weight (g/mol) 
  • (9)
  • (3)
Physical Form 
  • (9)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
Grade 
  • (4)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (9)
  • (9)
  • (3)
  • (4)
  • (9)
  • (3)
  • (9)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (5)