CAS RN 315704-66-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ BMI-1 Expression Inhibitor, PTC-209
A cell-permeable compound that downregulates cellular BMI-1 levels by inhibiting BMI-1 transcription.